missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Difluorophenyl isocyanate, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC308150050

 View more versions of this product

Catalog No. AC308150050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥96.0%
Liquid or Low Melting Solid
White to light yellow
96.0
Authentic
2, 6-Difluorophenyl isocyanate
MFCD00001998
97%
155.1
5g
100.0
Glass bottle
1.4945 to 1.4965 (20°C, 589nm)
65295-69-4
FC6H3NCO
C7H3F2NO

Chemical Identifiers

65295-69-4
100.0
C7H3F2NO
155.1
2, 6-Difluorophenyl isocyanate
96.0
97%
FC6H3NCO
MFCD00001998
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning