Please login to your online account to display your discounted pricing

2,6-Difluorophenyl isocyanate, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  308150010

 View more versions of this product

Catalog No. AC308150010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid or Low Melting Solid
Formula Weight 155.1
Quantity 1g
Color White to light yellow
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 2, 6-Difluorophenyl isocyanate
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C7H3F2NO
Linear Formula FC6H3NCO
MDL Number MFCD00001998
CAS 65295-69-4
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Flammable liquid and vapor.
Packaging Glass bottle
Refractive Index 1.4945 to 1.4965 (20°C, 589nm)
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2, 6-Difluorophenyl isocyanate
CAS 65295-69-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H3F2NO
Linear Formula FC6H3NCO
Formula Weight 155.1
MDL Number MFCD00001998
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Flammable liquid and vapor.

SDS & Safety Documents