Please login to your online account to display your discounted pricing

2,6-Difluorophenol, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  311630050

 View more versions of this product

Catalog No. AC311630050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Low Melting Solid
Formula Weight 130.1
Quantity 5g
Color Colorless
Melting Point 38° to 42°C
CAS Max % 100.0
Chemical Name or Material 2, 6-Difluorophenol
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C6H4F2O
Linear Formula FC6H3OH
MDL Number MFCD00002158
CAS 28177-48-2
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Flammable solid.
Packaging Glass bottle

Specifications

Chemical Name or Material 2, 6-Difluorophenol
CAS 28177-48-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H4F2O
Linear Formula FC6H3OH
Formula Weight 130.1
MDL Number MFCD00002158
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Flammable solid.

SDS & Safety Documents