Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Difluorophenol, 98+%

Chemical Identifiers

Chemical Name or Material 2,6-Difluorophenol
CAS 28177-48-2
Molecular Formula C6H4F2O
Formula Weight 130.09
MDL Number MFCD00002158
UN Number UN2921
Beilstein 2043613
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1603406

  SDS
alfa aesar™
A1603406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1603414

  SDS
alfa aesar™
A1603414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1603422

  SDS
alfa aesar™
A1603422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 130.09
Chemical Name or Material 2,6-Difluorophenol
Assay ≥98%
Molecular Formula C6H4F2O
MDL Number MFCD00002158
CAS 28177-48-2
UN Number UN2921
Beilstein 2043613
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H314-H318-H228
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, FLAMMABLE, N.O.S.
Flash Point 58°C (136°F)
Boiling Point 59° to 61°C (17mmHg)
Melting Point 36° to 40°C
Quantity 5g

Applications
2,6-Difluorophenol undergoes oxidative polymerization in the presence of the Fe-N,N′-bis(salicylidene)ethylenediamine (salen) complex (catalyst) and hydrogen peroxide (oxidizing agent) to give poly(2,6-difluoro-1,4-phenylene oxide). It is also used as pharmaceutical intermediates.

Solubility
Slightly soluble in water. Soluble in ethanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H314-H318-H228
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.