Please login to your online account to display your discounted pricing

2,6-Difluoronitrobenzene, 98%, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  AC397340010

 View more versions of this product

Catalog No. AC397340010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material 2, 6-Difluoronitrobenzene
Quantity 1g
CAS 19064-24-5
EINECS Number 242-793-8
Assay Percent Range 98%
Molecular Formula C6H3F2NO2
Linear Formula F2C6H3NO2
Formula Weight 159.09
MDL Number MFCD00192035

Specifications

Chemical Name or Material 2, 6-Difluoronitrobenzene
Assay Percent Range 98%
Molecular Formula C6H3F2NO2
Linear Formula F2C6H3NO2
Formula Weight 159.09
MDL Number MFCD00192035