Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Difluoromandelic acid, 97%

$63.11 - $158.54

Chemical Identifiers

Chemical Name or Material 2,6-Difluoromandelic acid
CAS 207981-50-8
Molecular Formula C8H6F2O3
Formula Weight 188.13
MDL Number MFCD00061298
Synonym alpha-Hydroxy-2,6-difluorophenylacetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1277303

 
alfa aesar™
L1277303
1g Each for $158.54

AAL12773MD

 
alfa aesar™
L12773MD
250mg Each for $63.11
Description & Specifications

Specifications

Formula Weight 188.13
Chemical Name or Material 2,6-Difluoromandelic acid
Assay 97%
Molecular Formula C8H6F2O3
MDL Number MFCD00061298
CAS 207981-50-8
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym alpha-Hydroxy-2,6-difluorophenylacetic acid
Health Hazard 1 Warning
Melting Point 142° to 145°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.