missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Difluoromandelic acid, 97%

Manufacturer:  Alfa Aesar™ L12773MD

 View more versions of this product

Catalog No. AAL12773MD



Specifications

Specifications

2,6-Difluoromandelic acid
250mg
C8H6F2O3
2,6-difluoromandelic acid, 2-2,6-difluorophenyl-2-hydroxyacetic acid, 2,6-difluorophenyl hydroxy acetic acid, 2,6-difluoromandelicacid, 2,6-difluorophenyl hydroxy acetic acid #, 2,6-difluoro-phenyl-hydroxy-acetic acid, 2-2,6-bis fluoranyl phenyl-2-oxidanyl-ethanoic acid
C1=CC(=C(C(=C1)F)C(C(=O)O)O)F
188.13
188.13
142°C to 145°C
207981-50-8
MFCD00061298
BMZWDQNFSCPFCG-UHFFFAOYSA-N
2-(2,6-difluorophenyl)-2-hydroxyacetic acid
588607
97%

Chemical Identifiers

207981-50-8
188.13
BMZWDQNFSCPFCG-UHFFFAOYSA-N
588607
C1=CC(=C(C(=C1)F)C(C(=O)O)O)F
C8H6F2O3
MFCD00061298
2,6-difluoromandelic acid, 2-2,6-difluorophenyl-2-hydroxyacetic acid, 2,6-difluorophenyl hydroxy acetic acid, 2,6-difluoromandelicacid, 2,6-difluorophenyl hydroxy acetic acid #, 2,6-difluoro-phenyl-hydroxy-acetic acid, 2-2,6-bis fluoranyl phenyl-2-oxidanyl-ethanoic acid
2-(2,6-difluorophenyl)-2-hydroxyacetic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents