Please login to your online account to display your discounted pricing

2,6-Difluorobenzyl chloride, 99%, Acros Organics

$228.93 - $228.93

Chemical Identifiers

Chemical Name or Material 2, 6-Difluorobenzyl chloride
Name Note 99%
CAS 697-73-4
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H5ClF2
Formula Weight 162.56
MDL Number MFCD00191346
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC346770050

  SDS
thermo fisher scientific
346770050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC346770250

  SDS
thermo fisher scientific
346770250
Glass bottle 25g Each for $228.93
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline
Formula Weight 162.56
Quantity 5g
Color Colorless
TSCA TSCA
CAS Max % 100.0
Synonym α-Chloro-2, 6-difluorotoluene
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 98.5
MDL Number MFCD00191346
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Harmful if swallowed.
Assay Percent Range 99%
Chemical Name or Material 2, 6-Difluorobenzyl chloride
Name Note 99%
CAS 697-73-4
Molecular Formula C7H5ClF2
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Harmful if swallowed.