Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Difluorobenzyl chloride, 98%

Chemical Identifiers

Chemical Name or Material 2,6-Difluorobenzyl chloride
CAS 697-73-4
Molecular Formula C7H5ClF2
Formula Weight 162.57
MDL Number MFCD00191346
UN Number UN1760
Beilstein 243793
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1947203

 
alfa aesar™
A1947203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1947206

 
alfa aesar™
A1947206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1947214

 
alfa aesar™
A1947214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 162.57
Chemical Name or Material 2,6-Difluorobenzyl chloride
Assay 98%
Molecular Formula C7H5ClF2
MDL Number MFCD00191346
CAS 697-73-4
UN Number UN1760
Beilstein 243793
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
Boiling Point 172° to 173°C (640mmHg)
Melting Point 32° to 35°C
Quantity 1g

Applications
2,6-Difluorobenzyl chloride is used as rufinamide intermediate. It is an important raw material and intermediate used in organic synthesis.

Solubility
Soluble in acetone, DMSO and DMF. Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.