Please login to your online account to display your discounted pricing

2,6-Difluorobenzoyl chloride, 99%, ACROS Organics™


Quantity 25mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  224510250

 View more versions of this product

Catalog No. AC224510250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 176.55
Quantity 25mL
Color Clear colorless to light yellow
Density 1.4g/mL
CAS Max % 100.0
Molecular Formula C7H3ClF2O
Chemical Name or Material 2, 6-Difluorobenzoyl chloride
Assay Percent Range 99%
CAS 18063-02-0
Health Hazard 1 GHS Signal Word: Danger
MDL Number MFCD00000659
CAS Min % 98.5
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4988 to 1.5008 (20°C, 589nm)
Linear Formula F2C6H3COCl
Health Hazard 2 GHS P Statement
Flammable liquid and vapor.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2, 6-Difluorobenzoyl chloride
CAS 18063-02-0
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H3ClF2O
Linear Formula F2C6H3COCl
Formula Weight 176.55
MDL Number MFCD00000659
Accessories

GHS Signal Word: Danger

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Flammable liquid and vapor.
Causes severe skin burns and eye damage.

SDS & Safety Documents