Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Difluorobenzophenone, 98%

$268.20 - $268.20

Chemical Identifiers

Chemical Name or Material 2,6-Difluorobenzophenone
CAS 59189-51-4
Molecular Formula C13H8F2O
Formula Weight 218.2
MDL Number MFCD00009895
Beilstein 2266589
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2273814

  SDS
alfa aesar™
B2273814
25g Each for $268.20
Description & Specifications

Specifications

Formula Weight 218.2
Chemical Name or Material 2,6-Difluorobenzophenone
Assay 98%
Molecular Formula C13H8F2O
MDL Number MFCD00009895
CAS 59189-51-4
Beilstein 2266589
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point >110°C (230°F)
Boiling Point 119° to 120°C (1.5mmHg)
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.