Please login to your online account to display your discounted pricing

2,6-Difluorobenzoic acid, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 6-Difluorobenzoic acid
Name Note 98%
CAS 385-00-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H4F2O2
Linear Formula F2C6H3CO2H
Formula Weight 158.1
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC183630250

  SDS
thermo fisher scientific
183630250
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder or Crystals
Formula Weight 158.1
Quantity 25g
Color Beige
EINECS Number 206-856-3
TSCA TSCA
RTECS Number DG8559000
Melting Point 156° to 160°C
CAS Max % 100.0
CAS Min % 97.5
Infrared Spectrum Authentic
Chemical Name or Material 2, 6-Difluorobenzoic acid
Name Note 98%
Assay Percent Range 98%
Molecular Formula C7H4F2O2
Linear Formula F2C6H3CO2H
MDL Number MFCD00002411
CAS 385-00-2
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.