missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Difluorobenzoic acid, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 183630250

 View more versions of this product

Catalog No. AC183630250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5%
158.1
Beige
100.0
97.5
F2C6H3CO2H
385-00-2
2, 6-Difluorobenzoic acid
Crystalline Powder or Crystals
25g
156° to 160°C
Authentic
MFCD00002411
98%
98%
C7H4F2O2

Chemical Identifiers

385-00-2
100.0
C7H4F2O2
158.1
2, 6-Difluorobenzoic acid, 98%
97.5
98%
F2C6H3CO2H
MFCD00002411
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 206-856-3

RTECSNumber : DG8559000

TSCA : TSCA