missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Difluorobenzoic acid, 98+%

Manufacturer:  Alfa Aesar™ A1455836

 View more versions of this product

Catalog No. AAA1455836



Specifications

Specifications

2,6-Difluorobenzoic acid
72°C to 77°C (13mmHg)
500g
C7H4F2O2
973774
ONOTYLMNTZNAQZ-UHFFFAOYSA-N
2,6-difluorobenzoic acid
9796
≥98%
Odorless
156°C to 160°C
385-00-2
MFCD00002411
benzoic acid, 2,6-difluoro, 2,6-difluorobenzoicacid, 2,6-difluoro benzoic acid, 2,6-difluoro-benzoic acid, pubchem2274, benzoic acid,6-difluoro, acmc-209j0h, 2,6,-difluorobenzoic acid, 3-09-00-01330 beilstein handbook reference, ksc223c7l
C1=CC(=C(C(=C1)F)C(=O)O)F
158.104
158.11

Chemical Identifiers

385-00-2
158.104
ONOTYLMNTZNAQZ-UHFFFAOYSA-N
9796
C1=CC(=C(C(=C1)F)C(=O)O)F
C7H4F2O2
MFCD00002411
benzoic acid, 2,6-difluoro, 2,6-difluorobenzoicacid, 2,6-difluoro benzoic acid, 2,6-difluoro-benzoic acid, pubchem2274, benzoic acid,6-difluoro, acmc-209j0h, 2,6,-difluorobenzoic acid, 3-09-00-01330 beilstein handbook reference, ksc223c7l
2,6-difluorobenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

RTECSNumber : DG8559000

Documents