Please login to your online account to display your discounted pricing

2,6-Difluorobenzoic acid, 97%, Maybridge


Quantity 1g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  SB01105DA

 View more versions of this product

Catalog No. SB01105DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 158.1
Quantity 1g
Assay 97%
CAS Min % 97.0
Packaging Amber glass bottle
CAS 385-00-2
Linear Formula F2C6H3CO2H
MDL Number MFCD00002411
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2, 6-Difluorobenzoic acid
Assay Percent Range 97%
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Molecular Formula C7H4F2O2

Specifications

Chemical Name or Material 2, 6-Difluorobenzoic acid
CAS 385-00-2
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H4F2O2
Linear Formula F2C6H3CO2H
Formula Weight 158.1
MDL Number MFCD00002411
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents