Please login to your online account to display your discounted pricing

2,6-Difluorobenzoic acid, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2, 6-Difluorobenzoic acid
CAS 385-00-2
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H4F2O2
Linear Formula F2C6H3CO2H
Formula Weight 158.1
MDL Number MFCD00002411
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SB01105DA

 
fisher scientific company
SB01105DA
Amber glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 158.1
Quantity 1g
Assay 97%
CAS Min % 97.0
Linear Formula F2C6H3CO2H
CAS 385-00-2
Packaging Amber glass bottle
Chemical Name or Material 2, 6-Difluorobenzoic acid
Assay Percent Range 97%
MDL Number MFCD00002411
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C7H4F2O2