Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Difluorobenzamide, 98+%

$803.11 - $803.11

Chemical Identifiers

Chemical Name or Material 2,6-Difluorobenzamide
CAS 18063-03-1
Molecular Formula C7H5F2NO
Formula Weight 157.12
MDL Number MFCD00007972
Beilstein 2047480
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1168536

  SDS
alfa aesar™
A1168536
500g Each for $803.11
Description & Specifications

Specifications

Formula Weight 157.12
Assay ≥98%
MDL Number MFCD00007972
Melting Point 145° to 148°C
Quantity 500g
Chemical Name or Material 2,6-Difluorobenzamide
Beilstein 2047480
CAS 18063-03-1
Molecular Formula C7H5F2NO
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H332-H315-H319-H335
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H332-H315-H319-H335
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.