Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Difluoroaniline, 98%

Chemical Identifiers

Chemical Name or Material 2,6-Difluoroaniline
CAS 5509-65-9
Molecular Formula C6H5F2N
Formula Weight 129.11
MDL Number MFCD00007655
UN Number UN1992
Beilstein 2802697
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1302014

  SDS
alfa aesar™
A1302014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1302022

  SDS
alfa aesar™
A1302022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 129.11
Chemical Name or Material 2,6-Difluoroaniline
UN Number UN1992
Health Hazard 1 Danger
Boiling Point 152° to 154°C
Health Hazard 2 GHS H Statement
H311-H331-H226-H373-H302-H315-H319
Toxic in contact with skin.
Toxic if inhaled.
Flammable liquid and vapour.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Beilstein 2802697
MDL Number MFCD00007655
Molecular Formula C6H5F2N
Quantity 25g
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Flash Point 51°C (124°F)
CAS 5509-65-9
Assay 98%
Odor Characteristic
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.

Applications
2,6-Difluoroaniline is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Incompatible to acid chlorides, acid anhydrides, oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

GHS H Statement
H311-H331-H226-H373-H302-H315-H319
Toxic in contact with skin.
Toxic if inhaled.
Flammable liquid and vapour.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.