missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Difluoro-4-methoxyphenol, 97%

$204.95 - $822.76

Chemical Identifiers

CAS 886498-93-7
Molecular Formula C7H6F2O2
Formula Weight 160.12
MDL Number MFCD04115926
Chemical Name or Material 2,6-Difluoro-4-methoxyphenol
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2667303 SDS Alfa Aesar™
H2667303
1g Each for $204.95
AAH2667306 SDS Alfa Aesar™
H2667306
5g Each for $822.76
Specifications Description & Specifications

Chemical Identifiers

886498-93-7
160.12
2,6-Difluoro-4-methoxyphenol
C7H6F2O2
MFCD04115926

Specifications

67° to 71°C
886498-93-7
160.12
2,6-Difluoro-4-methoxyphenol
1g
C7H6F2O2
MFCD04115926
97%

Safety and Handling

GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P280-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention.

Danger