Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Difluoro-4-methoxyaniline, 97%

Chemical Identifiers

Chemical Name or Material 2,6-Difluoro-4-methoxyaniline
CAS 151414-47-0
Molecular Formula C7H7F2NO
Formula Weight 159.14
MDL Number MFCD04115922
UN Number UN2811
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2672803

  SDS
alfa aesar™
H2672803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2672806

  SDS
alfa aesar™
H2672806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 159.14
MDL Number MFCD04115922
Molecular Formula C7H7F2NO
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Assay 97%
Quantity 1g
UN Number UN2811
Health Hazard 1 Danger
CAS 151414-47-0
Melting Point 38° to 40°C
Health Hazard 2 GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Chemical Name or Material 2,6-Difluoro-4-methoxyaniline

Applications
2,6-Difluoro-4-methoxyaniline is used as primary and secondary intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.