missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Difluoro-3-nitrotoluene, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H63431MD

 View more versions of this product

Catalog No. AAH63431MD


Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2,6-Difluoro-3-nitrotoluene used as a intermediate in organic synthesis, pharmaceutical and chemical research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Specifications

2,6-Difluoro-3-nitrotoluene
C7H5F2NO2
250 mg
2,6-difluoro-3-nitrotoluene, benzene, 1,3-difluoro-2-methyl-4-nitro, 2,4-difluoro-3-methylnitrobenzene, pubchem10410, pubchem15439, 1,3-difluoro-2-methyl-4-nitro-benzene, 1,3-bis fluoranyl-2-methyl-4-nitro-benzene
AVWNNVJXXMKAPB-UHFFFAOYSA-N
1,3-difluoro-2-methyl-4-nitrobenzene
12795486
98%
79562-49-5
MFCD11519030
UN2811
Slightly soluble in water.
CC1=C(C=CC(=C1F)[N+](=O)[O-])F
173.119
173.12
Solid

Chemical Identifiers

79562-49-5
173.119
AVWNNVJXXMKAPB-UHFFFAOYSA-N
12795486
CC1=C(C=CC(=C1F)[N+](=O)[O-])F
C7H5F2NO2
MFCD11519030
2,6-difluoro-3-nitrotoluene, benzene, 1,3-difluoro-2-methyl-4-nitro, 2,4-difluoro-3-methylnitrobenzene, pubchem10410, pubchem15439, 1,3-difluoro-2-methyl-4-nitro-benzene, 1,3-bis fluoranyl-2-methyl-4-nitro-benzene
1,3-difluoro-2-methyl-4-nitrobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions