missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Difluoro-3-nitrobenzeneboronic acid, 96%

$70.43 - $835.59

Chemical Identifiers

CAS 1150114-28-5
Molecular Formula C6H4BF2NO4
Molecular Weight (g/mol) 202.908
MDL Number MFCD12025957
InChI Key SPEDPBJIXAVPMJ-UHFFFAOYSA-N
Synonym 2,6-difluoro-3-nitro-phenyl boronic acid, 2,6-difluoro-3-nitrophenyl boronic acid
PubChem CID 46738911
IUPAC Name (2,6-difluoro-3-nitrophenyl)boronic acid
SMILES B(C1=C(C=CC(=C1F)[N+](=O)[O-])F)(O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH61530MD
View Documents
Alfa Aesar™
H61530MD
250mg Each for $70.43
 
AAH6153003
View Documents
Alfa Aesar™
H6153003
1g Each for $208.49
 
AAH6153006
View Documents
Alfa Aesar™
H6153006
5g Each for $835.59
 
Specifications

Chemical Identifiers

1150114-28-5
202.908
SPEDPBJIXAVPMJ-UHFFFAOYSA-N
46738911
B(C1=C(C=CC(=C1F)[N+](=O)[O-])F)(O)O
C6H4BF2NO4
MFCD12025957
2,6-difluoro-3-nitro-phenyl boronic acid, 2,6-difluoro-3-nitrophenyl boronic acid
(2,6-difluoro-3-nitrophenyl)boronic acid

Specifications

250mg
C6H4BF2NO4
2,6-difluoro-3-nitro-phenyl boronic acid, 2,6-difluoro-3-nitrophenyl boronic acid
B(C1=C(C=CC(=C1F)[N+](=O)[O-])F)(O)O
202.908
202.19
2,6-Difluoro-3-nitrobenzeneboronic acid
1150114-28-5
MFCD12025957
SPEDPBJIXAVPMJ-UHFFFAOYSA-N
(2,6-difluoro-3-nitrophenyl)boronic acid
46738911
96%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures