missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Difluoro-3-nitrobenzeneboronic acid, 96%

Manufacturer:  Alfa Aesar™ H6153006

 View more versions of this product

Catalog No. AAH6153006



Specifications

Specifications

2,6-Difluoro-3-nitrobenzeneboronic acid
1150114-28-5
MFCD12025957
SPEDPBJIXAVPMJ-UHFFFAOYSA-N
(2,6-difluoro-3-nitrophenyl)boronic acid
46738911
96%
5g
C6H4BF2NO4
2,6-difluoro-3-nitro-phenyl boronic acid, 2,6-difluoro-3-nitrophenyl boronic acid
B(C1=C(C=CC(=C1F)[N+](=O)[O-])F)(O)O
202.908
202.19

Chemical Identifiers

1150114-28-5
202.908
SPEDPBJIXAVPMJ-UHFFFAOYSA-N
46738911
B(C1=C(C=CC(=C1F)[N+](=O)[O-])F)(O)O
C6H4BF2NO4
MFCD12025957
2,6-difluoro-3-nitro-phenyl boronic acid, 2,6-difluoro-3-nitrophenyl boronic acid
(2,6-difluoro-3-nitrophenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents