Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Difluoro-3-methylphenol, 97+%

Chemical Identifiers

Chemical Name or Material 2,6-Difluoro-3-methylphenol
CAS 261763-46-6
Molecular Formula C7H6F2O
Formula Weight 144.12
MDL Number MFCD01631340
UN Number UN3265
Synonym 2,6-Difluoro-m-cresol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2408203

  SDS
alfa aesar™
B2408203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2408206

  SDS
alfa aesar™
B2408206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 144.12
Assay ≥97%
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
MDL Number MFCD01631340
Boiling Point 182°C
Quantity 1g
UN Number UN3265
Health Hazard 2 GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Synonym 2,6-Difluoro-m-cresol
Health Hazard 1 Danger
Molecular Formula C7H6F2O
Chemical Name or Material 2,6-Difluoro-3-methylphenol
CAS 261763-46-6
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.