missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Difluoro-3-methylbenzoyl chloride, 97%

$80.04 - $276.04

Chemical Identifiers

CAS 261763-39-7
Molecular Formula C8H5ClF2O
Molecular Weight (g/mol) 190.574
MDL Number MFCD01631334
InChI Key LXCIIPPXRHOCQN-UHFFFAOYSA-N
Synonym 2,6-difluoro-3-methyl-benzoyl chloride, 2,6-difluoro-3-methylbenzoylchloride, acmc-20ao8t, 2,6-difluoro-m-toluoyl chloride, 2,6-difluoro-3-methyl benzoyl chloride, 2,6-difluoro-m-toluoyl chloride, benzoyl chloride,2,6-difluoro-3-methyl, benzoyl chloride, 2,6-difluoro-3-methyl, 2,6-bis fluoranyl-3-methyl-benzoyl chloride, 2,6-difluoro-3-methylbenzoyl chloride
PubChem CID 2774145
IUPAC Name 2,6-difluoro-3-methylbenzoyl chloride
SMILES CC1=C(C(=C(C=C1)F)C(=O)Cl)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1815903 View Documents Alfa Aesar™
L1815903
1g Each for $80.04
 
AAL1815906 View Documents Alfa Aesar™
L1815906
5g Each for $276.04
 
Specifications

Chemical Identifiers

261763-39-7
190.574
LXCIIPPXRHOCQN-UHFFFAOYSA-N
2774145
CC1=C(C(=C(C=C1)F)C(=O)Cl)F
C8H5ClF2O
MFCD01631334
2,6-difluoro-3-methyl-benzoyl chloride, 2,6-difluoro-3-methylbenzoylchloride, acmc-20ao8t, 2,6-difluoro-m-toluoyl chloride, 2,6-difluoro-3-methyl benzoyl chloride, 2,6-difluoro-m-toluoyl chloride, benzoyl chloride,2,6-difluoro-3-methyl, benzoyl chloride, 2,6-difluoro-3-methyl, 2,6-bis fluoranyl-3-methyl-benzoyl chloride, 2,6-difluoro-3-methylbenzoyl chloride
2,6-difluoro-3-methylbenzoyl chloride

Specifications

1g
C8H5ClF2O
UN3265
LXCIIPPXRHOCQN-UHFFFAOYSA-N
2,6-difluoro-3-methylbenzoyl chloride
2774145
97%
261763-39-7
MFCD01631334
2,6-difluoro-3-methyl-benzoyl chloride, 2,6-difluoro-3-methylbenzoylchloride, acmc-20ao8t, 2,6-difluoro-m-toluoyl chloride, 2,6-difluoro-3-methyl benzoyl chloride, 2,6-difluoro-m-toluoyl chloride, benzoyl chloride,2,6-difluoro-3-methyl, benzoyl chloride, 2,6-difluoro-3-methyl, 2,6-bis fluoranyl-3-methyl-benzoyl chloride, 2,6-difluoro-3-methylbenzoyl chloride
CC1=C(C(=C(C=C1)F)C(=O)Cl)F
190.574
190.58
2,6-Difluoro-3-methylbenzoyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.