Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Difluoro-3-methylbenzoic acid, 97+%

$50.95 - $172.41

Chemical Identifiers

Chemical Name or Material 2,6-Difluoro-3-methylbenzoic acid
CAS 32890-88-3
Molecular Formula C8H6F2O2
Formula Weight 172.13
MDL Number MFCD01631332
Synonym 2,6-Difluoro-m-toluic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2352503

 
alfa aesar™
B2352503
1g Each for $50.95

AAB2352506

 
alfa aesar™
B2352506
5g Each for $172.41
Description & Specifications

Specifications

Formula Weight 172.13
Chemical Name or Material 2,6-Difluoro-3-methylbenzoic acid
Assay ≥97%
Molecular Formula C8H6F2O2
MDL Number MFCD01631332
CAS 32890-88-3
Synonym 2,6-Difluoro-m-toluic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 138° to 142°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.