Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Difluoro-3-methylbenzaldehyde, 97+%

$71.27 - $241.08

Chemical Identifiers

Chemical Name or Material 2,6-Difluoro-3-methylbenzaldehyde
CAS 261763-34-2
Molecular Formula C8H6F2O
Formula Weight 156.13
MDL Number MFCD01631333
Synonym 2,6-Difluoro-m-tolualdehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2419703

  SDS
alfa aesar™
B2419703
1g Each for $71.27

AAB2419706

  SDS
alfa aesar™
B2419706
5g Each for $241.08
Description & Specifications

Specifications

Formula Weight 156.13
Chemical Name or Material 2,6-Difluoro-3-methylbenzaldehyde
Assay ≥97%
Molecular Formula C8H6F2O
MDL Number MFCD01631333
CAS 261763-34-2
Synonym 2,6-Difluoro-m-tolualdehyde
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.