Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Difluoro-3-methylanisole, 97%

$78.81 - $237.05

Chemical Identifiers

Chemical Name or Material 2,6-Difluoro-3-methylanisole
CAS 261763-33-1
Molecular Formula C8H8F2O
Formula Weight 158.15
MDL Number MFCD01631341
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2672203

 
alfa aesar™
H2672203
1g Each for $78.81

AAH2672206

 
alfa aesar™
H2672206
5g Each for $237.05
Description & Specifications

Specifications

Formula Weight 158.15
Chemical Name or Material 2,6-Difluoro-3-methylanisole
Assay 97%
Molecular Formula C8H8F2O
MDL Number MFCD01631341
CAS 261763-33-1
Boiling Point 53°C (1mmHg)
Quantity 1g