Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Difluoro-3-methoxyphenol, 97%

$144.69 - $560.30

Chemical Identifiers

Chemical Name or Material 2,6-Difluoro-3-methoxyphenol
CAS 886498-60-8
Molecular Formula C7H6F2O2
Formula Weight 160.12
MDL Number MFCD04115907
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2681203

  SDS
alfa aesar™
H2681203
1g Each for $144.69

AAH2681206

  SDS
alfa aesar™
H2681206
5g Each for $560.30
Description & Specifications

Specifications

Formula Weight 160.12
Chemical Name or Material 2,6-Difluoro-3-methoxyphenol
Assay 97%
Molecular Formula C7H6F2O2
MDL Number MFCD04115907
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Quantity 1g
CAS 886498-60-8
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention.
Melting Point 73° to 76°C
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention.

GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.