Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Difluoro-3-methoxybenzoic acid, 97%

$105.03 - $575.58

Chemical Identifiers

Chemical Name or Material 2,6-Difluoro-3-methoxybenzoic acid
CAS 886498-30-2
Molecular Formula C8H6F2O3
Formula Weight 188.13
MDL Number MFCD04115901
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2671203

  SDS
alfa aesar™
H2671203
1g Each for $105.03

AAH2671209

  SDS
alfa aesar™
H2671209
10g Each for $575.58
Description & Specifications

Specifications

Formula Weight 188.13
CAS 886498-30-2
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2,6-Difluoro-3-methoxybenzoic acid
Assay 97%
Molecular Formula C8H6F2O3
MDL Number MFCD04115901
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g

Applications
As pharmaceuticals, they find applications in antibiotics, as sedatives, and for cancer treatment. Used frequently in organic synthesis. Find application as propellants.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.