Learn More
2,6-Difluoro-3-methoxyaniline, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2663103
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2,6-Difluoro-3-methoxyaniline | |
C7H7F2NO | |
MFCD04115898 | |
UN2810 | |
XVLHUQRLEPQGDI-UHFFFAOYSA-N | |
2,6-difluoro-3-methoxyaniline | |
3871278 | |
97% |
144851-62-7 | |
1.5235 | |
1 g | |
benzenamine,2,6-difluoro-3-methoxy, acmc-20dww5, 2,6-difluoro-m-anisidine, 3-amino-2,4-difluoroanisole, 2,6-difluoro-3-methoxyphenylamine, benzenamine, 2,6-difluoro-3-methoxy-9ci | |
COC1=C(F)C(N)=C(F)C=C1 | |
159.14 | |
159.14 |
Chemical Identifiers
144851-62-7 | |
159.14 | |
XVLHUQRLEPQGDI-UHFFFAOYSA-N | |
3871278 | |
COC1=C(F)C(N)=C(F)C=C1 |
C7H7F2NO | |
MFCD04115898 | |
benzenamine,2,6-difluoro-3-methoxy, acmc-20dww5, 2,6-difluoro-m-anisidine, 3-amino-2,4-difluoroanisole, 2,6-difluoro-3-methoxyphenylamine, benzenamine, 2,6-difluoro-3-methoxy-9ci | |
2,6-difluoro-3-methoxyaniline |
Safety and Handling
GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
H302+H332-H311-H315-H319
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only