missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,6-Dichlorotoluene, 99+%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC113790025
Specifications
2, 6-Dichlorotoluene | |
82°C | |
Glass bottle | |
1.25 | |
Colorless to Yellow | |
2.5L | |
118-69-4 | |
100.0 | |
C7H6Cl2 | |
MFCD00000576 | |
1,3-dichloro-2-methyl-benzene, 2,6-dichloro toluene, 2,6-dichloro-toluene, 2,6-dichlorotoluene, benzene, 1,3-dichloro-2-methyl, benzene,dichloromethyl, pubchem3701, toluene, 2,6-dichloro, toluene,6-dichloro, unii-rvm09757ww | |
DMEDNTFWIHCBRK-UHFFFAOYSA-N | |
1,3-dichloro-2-methylbenzene | |
8368 | |
Liquid |
1.2500g/mL | |
Authentic | |
1.5500 to 1.552 | |
196°C to 203°C | |
2°C | |
99+% | |
99.0 | |
99% min. (GC) | |
CH3C6H3Cl2 | |
05, 296 | |
Solubility in water: 24mg/l (20°C). Other solubilities: soluble in alcohol,ether and acetone | |
CC1=C(C=CC=C1Cl)Cl | |
161.03 | |
161.03 | |
99+% |
Chemical Identifiers
118-69-4 | |
161.03 | |
DMEDNTFWIHCBRK-UHFFFAOYSA-N | |
8368 | |
CC1=C(C=CC=C1Cl)Cl |
C7H6Cl2 | |
MFCD00000576 | |
1,3-dichloro-2-methyl-benzene, 2,6-dichloro toluene, 2,6-dichloro-toluene, 2,6-dichlorotoluene, benzene, 1,3-dichloro-2-methyl, benzene,dichloromethyl, pubchem3701, toluene, 2,6-dichloro, toluene,6-dichloro, unii-rvm09757ww | |
1,3-dichloro-2-methylbenzene |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 204-269-7
TSCA : TSCA