missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,6-Dichlorothiobenzamide, 97%, Thermo Scientific™
$199.47 - $199.47
Chemical Identifiers
CAS | 1918-13-4 |
---|---|
Molecular Formula | C7H5Cl2NS |
Molecular Weight (g/mol) | 206.08 |
MDL Number | MFCD00053523 |
InChI Key | KGKGSIUWJCAFPX-UHFFFAOYSA-N |
Synonym | 2,6-dichlorothiobenzamide, chlorthiamid, chlorthiamide, chlorothiamide, chlorthioamide, chlortiamid, prefix, dcbn, benzenecarbothioamide, 2,6-dichloro, 2,6-dichlorobenzothioamide |
PubChem CID | 2734819 |
ChEBI | CHEBI:949 |
SMILES | NC(=S)C1=C(Cl)C=CC=C1Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH2663603
|
Thermo Scientific Chemicals
H2663603 |
1 g |
Each for $199.47
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1918-13-4 | |
206.08 | |
KGKGSIUWJCAFPX-UHFFFAOYSA-N | |
2734819 | |
NC(=S)C1=C(Cl)C=CC=C1Cl |
C7H5Cl2NS | |
MFCD00053523 | |
2,6-dichlorothiobenzamide, chlorthiamid, chlorthiamide, chlorothiamide, chlorthioamide, chlortiamid, prefix, dcbn, benzenecarbothioamide, 2,6-dichloro, 2,6-dichlorobenzothioamide | |
CHEBI:949 |
Specifications
1918-13-4 | |
>100°C (212°F) | |
MFCD00053523 | |
1910353 | |
KGKGSIUWJCAFPX-UHFFFAOYSA-N | |
2,6-dichlorobenzene-1-carbothioamide | |
2734819 | |
206.09 | |
2,6-Dichlorothiobenzamide |
149°C to 153°C | |
C7H5Cl2NS | |
1 g | |
2,6-dichlorothiobenzamide, chlorthiamid, chlorthiamide, chlorothiamide, chlorthioamide, chlortiamid, prefix, dcbn, benzenecarbothioamide, 2,6-dichloro, 2,6-dichlorobenzothioamide | |
NC(=S)C1=C(Cl)C=CC=C1Cl | |
206.08 | |
CHEBI:949 | |
97% |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 217-637-7
RTECSNumber : CV3850000
TSCA : No
Recommended Storage : Ambient temperatures