missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Dichloropyrazine, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 232540050

 View more versions of this product

Catalog No. AC232540050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2, 6-Dichloropyrazine
Authentic
White to Beige
5g
99.0
99% min. (GC)
23, III, 903
LSEAAPGIZCDEEH-UHFFFAOYSA-N
2,6-dichloropyrazine
78504
Crystals or Crystalline Powder
>110°C
Glass bottle
53.0°C to 56.0°C
4774-14-5
100.0
C4H2Cl2N2
2, 6-dichloropyrazine, 2,6 dichloropyrazine, 2,6-chloro pyrazine, 2,6-dichloro pyrazine, 2,6-dichloro-pyrazin, 2,6-dichloro-pyrazine, 2,6-dicloropyrazine, 2-chloro-6-chloro-pyrazine, pubchem8548, pyrazine, 2,6-dichloro
C1=C(N=C(C=N1)Cl)Cl
148.98
148.98
99+%

Chemical Identifiers

4774-14-5
148.98
2, 6-dichloropyrazine, 2,6 dichloropyrazine, 2,6-chloro pyrazine, 2,6-dichloro pyrazine, 2,6-dichloro-pyrazin, 2,6-dichloro-pyrazine, 2,6-dicloropyrazine, 2-chloro-6-chloro-pyrazine, pubchem8548, pyrazine, 2,6-dichloro
2,6-dichloropyrazine
C4H2Cl2N2
LSEAAPGIZCDEEH-UHFFFAOYSA-N
78504
C1=C(N=C(C=N1)Cl)Cl
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

EINECSNumber : 225-316-8

SDS Documents