missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Dichlorophenylboronic acid, 97%, Maybridge

Manufacturer:  Fisher Scientific Company AC35911DA

 View more versions of this product

Catalog No. AC35911DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2, 6-Dichlorophenylboronic acid
Amber Glass Bottle
1g
97.5
98%
MFCD00064869
CXDPUSMFYPQXCV-UHFFFAOYSA-N
(2,6-dichlorophenyl)boronic acid
2734332
Powder
Authentic
White
73852-17-2
100.0
C6H5BCl2O2
2,6-dichloro-phenylboronic acid, 2,6-dichlorobenzeneboronic acid, 2,6-dichlorophenyl boranediol, 2,6-dichlorophenyl boronic acid, 2,6-dichlorophenylboronicacid, acmc-1bc3m, boronic acid, 2,6-dichlorophenyl, ksc489k8l, pubchem7938
B(C1=C(C=CC=C1Cl)Cl)(O)O
190.814
190.82
≥97.5%

Chemical Identifiers

73852-17-2
190.814
CXDPUSMFYPQXCV-UHFFFAOYSA-N
2734332
B(C1=C(C=CC=C1Cl)Cl)(O)O
C6H5BCl2O2
MFCD00064869
2,6-dichloro-phenylboronic acid, 2,6-dichlorobenzeneboronic acid, 2,6-dichlorophenyl boranediol, 2,6-dichlorophenyl boronic acid, 2,6-dichlorophenylboronicacid, acmc-1bc3m, boronic acid, 2,6-dichlorophenyl, ksc489k8l, pubchem7938
(2,6-dichlorophenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents