Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dichlorophenylacetonitrile, 98%

Chemical Identifiers

Chemical Name or Material 2,6-Dichlorophenylacetonitrile
CAS 3215-64-3
Molecular Formula C8H5Cl2N
Formula Weight 186.04
MDL Number MFCD00001901
UN Number UN3439
Beilstein 1910482
Synonym 2,6-Dichlorobenzyl cyanide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1998314

 
alfa aesar™
A1998314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1998322

 
alfa aesar™
A1998322
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1998336

 
alfa aesar™
A1998336
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 186.04
Chemical Name or Material 2,6-Dichlorophenylacetonitrile
Odor Characteristic
Assay 98%
Molecular Formula C8H5Cl2N
MDL Number MFCD00001901
CAS 3215-64-3
UN Number UN3439
Beilstein 1910482
Melting Point 74° to 77°C
Synonym 2,6-Dichlorobenzyl cyanide
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H331-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Quantity 25g
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301-H311-H331-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.