missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Dichlorophenylacetonitrile 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 155081000

 View more versions of this product

Catalog No. AC155081000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2, 6-Dichlorophenylacetonitrile
74° to 77°C
97%
96.0
Authentic
≥96.0%
C8H5Cl2N
186.04
Glass bottle
White to light yellow
100g
3215-64-3
100.0
Crystalline Powder
97%
Cl2C6H3CH2CN
MFCD00001901
2, 6-Dichlorobenzyl cyanide

Chemical Identifiers

3215-64-3
100.0
C8H5Cl2N
186.04
2, 6-Dichlorobenzyl cyanide
≥96.0%
96.0
97%
Cl2C6H3CH2CN
MFCD00001901
2, 6-Dichlorophenylacetonitrile, 97%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 221-730-8

TSCA : TSCA