missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Dichlorophenylacetonitrile 97%, ACROS Organics™

Manufacturer:  Acros Organics 155080250

 View more versions of this product

Catalog No. AC155080250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2, 6-Dichlorophenylacetonitrile
74.0°C to 77.0°C
97%
96.0
Authentic
97%
Cl2C6H3CH2CN
Glass bottle
AOEJUUCUKRUCEF-UHFFFAOYSA-N
2-(2,6-dichlorophenyl)acetonitrile
76678
Crystalline Powder
White through Yellow
25g
3215-64-3
100.0
97%
C8H5Cl2N
MFCD00001901
2, 6-Dichlorobenzyl cyanide
C1=CC(=C(C(=C1)Cl)CC#N)Cl
186.035
186.04

Chemical Identifiers

3215-64-3
186.035
AOEJUUCUKRUCEF-UHFFFAOYSA-N
76678
C8H5Cl2N
MFCD00001901
2, 6-Dichlorobenzyl cyanide
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 221-730-8

TSCA : TSCA

SDS Documents