Please login to your online account to display your discounted pricing

2,6-Dichlorophenylacetonitrile 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 6-Dichlorophenylacetonitrile
Name Note 97%
CAS 3215-64-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H5Cl2N
Linear Formula Cl2C6H3CH2CN
Formula Weight 186.04
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC155080250

  SDS
thermo fisher scientific
155080250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC155081000

  SDS
thermo fisher scientific
155081000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 186.04
Quantity 25g
Color White to light yellow
EINECS Number 221-730-8
TSCA TSCA
Melting Point 74° to 77°C
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 2, 6-Dichlorophenylacetonitrile
Infrared Spectrum Authentic
Name Note 97%
Assay Percent Range 97%
Molecular Formula C8H5Cl2N
Linear Formula Cl2C6H3CH2CN
MDL Number MFCD00001901
Packaging Glass bottle
CAS 3215-64-3
Synonym 2, 6-Dichlorobenzyl cyanide
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.