Please login to your online account to display your discounted pricing

2,6-Dichlorophenylacetone, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 6-Dichlorophenylacetone
Name Note 98%
CAS 93457-06-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H8Cl2O
Formula Weight 203.07
MDL Number MFCD03410452
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC350820050

  SDS
thermo fisher scientific
35082-0050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 203.07
Physical Form Liquid After Melting
Quantity 5g
Color Clear light yellow
TSCA TSCA
CAS Max % 100.0
Refractive Index 1.5490 to 1.5510 (20°C, 589nm)
Infrared Spectrum Authentic
CAS Min % 97.5
Assay Percent Range 98%
Chemical Name or Material 2, 6-Dichlorophenylacetone
Name Note 98%
CAS 93457-06-8
Molecular Formula C9H8Cl2O
MDL Number MFCD03410452