Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dichlorophenylacetamide, 98+%

$112.76 - $391.54

Chemical Identifiers

Chemical Name or Material 2,6-Dichlorophenylacetamide
CAS 78433-88-2
Molecular Formula C8H7Cl2NO
Formula Weight 204.06g/mol
MDL Number MFCD00082745
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1288014

 
alfa aesar™
L12880-14
25g Each for $391.54

AAL1288006

 
alfa aesar™
L12880-06
5g Each for $112.76
Description & Specifications

Specifications

Melting Point 216° to 218°C
Quantity 25g
Chemical Name or Material 2,6-Dichlorophenylacetamide
CAS 78433-88-2
Assay ≥98%
Molecular Formula C8H7Cl2NO
Formula Weight 204.06g/mol
MDL Number MFCD00082745