Please login to your online account to display your discounted pricing

2,6-Dichlorophenyl isocyanate, 98%, ACROS Organics™


Quantity 5g
Packaging Glass Bottle in Tin Can

Manufacturer: thermo fisher scientific  33497-0050

 View more versions of this product

Catalog No. AC334970050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 188.01
Physical Form Crystalline Mass
Quantity 5g
Color White to yellow
Packaging Glass Bottle in Tin Can
Density 76g/mL
Melting Point 42° to 46°C
CAS Max % 100.0
Chemical Name or Material 2, 6-Dichlorophenyl isocyanate
Infrared Spectrum Authentic
Molecular Formula C7H3Cl2NO
Assay Percent Range 98%
CAS 39920-37-1
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00002003
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Toxic if swallowed.
Causes serious eye irritation.
Toxic in contact with skin.
Causes skin irritation.
Toxic if inhaled.
May cause respiratory irritation.

Specifications

Chemical Name or Material 2, 6-Dichlorophenyl isocyanate
CAS 39920-37-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H3Cl2NO
Formula Weight 188.01
MDL Number MFCD00002003
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Toxic if swallowed.
Causes serious eye irritation.
Toxic in contact with skin.
Causes skin irritation.
Toxic if inhaled.
May cause respiratory irritation.

SDS & Safety Documents