missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,6-Dichlorophenol, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 113591000
Specifications
2, 6-Dichlorophenol | |
Glass bottle | |
White to Yellow | |
100g | |
87-65-0 | |
100.0 | |
C6H4Cl2O | |
MFCD00002176 | |
15, 3083 | |
Solubility in water: 1.9g/L (25°C). Other solubilities: soluble in petroleum ether,benzene,diethyl ether,and ethanol | |
C1=CC(=C(C(=C1)Cl)O)Cl | |
163 | |
CHEBI:28457 | |
Crystals or Crystalline Powder |
Authentic | |
218.0°C to 220.0°C | |
65.0°C to 68.0°C | |
99% | |
98.5 | |
98.5% min. (GC) | |
Cl2C6H3OH | |
06, 190 | |
2,6 dichlorophenol, 2,6-dcp, 2,6-dichlorfenol, 2,6-dichlorfenol czech, 2,6-dichloro-phenol, 2,6-dichlorphenol, ccris 2511, phenol, 2,6-dichloro, rcra waste number u082, unii-q7e9k52w7e | |
HOLHYSJJBXSLMV-UHFFFAOYSA-N | |
2,6-dichlorophenol | |
6899 | |
163 | |
99% |
Chemical Identifiers
87-65-0 | |
163 | |
HOLHYSJJBXSLMV-UHFFFAOYSA-N | |
6899 | |
2,6-dichlorophenol |
C6H4Cl2O | |
MFCD00002176 | |
2,6 dichlorophenol, 2,6-dcp, 2,6-dichlorfenol, 2,6-dichlorfenol czech, 2,6-dichloro-phenol, 2,6-dichlorphenol, ccris 2511, phenol, 2,6-dichloro, rcra waste number u082, unii-q7e9k52w7e | |
CHEBI:28457 | |
C1=CC(=C(C(=C1)Cl)O)Cl |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Danger
EINECSNumber : 201-761-3
RTECSNumber : SK8750000
TSCA : TSCA