Learn More
2,6-Dichlorophenol, 99%, Thermo Scientific Chemicals
$132.44 - $446.60
Chemical Identifiers
CAS | 87-65-0 |
---|---|
Molecular Formula | C6H4Cl2O |
Molecular Weight (g/mol) | 163.00 |
MDL Number | MFCD00002176 |
InChI Key | HOLHYSJJBXSLMV-UHFFFAOYSA-N |
Synonym | phenol, 2,6-dichloro, 2,6-dichlorfenol, 2,6-dichlorphenol, rcra waste number u082, 2,6-dichloro-phenol, 2,6-dichlorfenol czech, unii-q7e9k52w7e, 2,6 dichlorophenol, ccris 2511, 2,6-dcp |
PubChem CID | 6899 |
ChEBI | CHEBI:28457 |
IUPAC Name | 2,6-dichlorophenol |
SMILES | OC1=C(Cl)C=CC=C1Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1441122
|
Thermo Scientific Chemicals
A1441122 |
100 g |
Each for $132.44
|
|
|||||
AAA1441136
|
Thermo Scientific Chemicals
A1441136 |
500 g |
Each for $446.60
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
87-65-0 | |
163.00 | |
HOLHYSJJBXSLMV-UHFFFAOYSA-N | |
6899 | |
2,6-dichlorophenol |
C6H4Cl2O | |
MFCD00002176 | |
phenol, 2,6-dichloro, 2,6-dichlorfenol, 2,6-dichlorphenol, rcra waste number u082, 2,6-dichloro-phenol, 2,6-dichlorfenol czech, unii-q7e9k52w7e, 2,6 dichlorophenol, ccris 2511, 2,6-dcp | |
CHEBI:28457 | |
OC1=C(Cl)C=CC=C1Cl |
Specifications
87-65-0 | |
218°C to 220°C | |
C6H4Cl2O | |
100 g | |
1447806 | |
phenol, 2,6-dichloro, 2,6-dichlorfenol, 2,6-dichlorphenol, rcra waste number u082, 2,6-dichloro-phenol, 2,6-dichlorfenol czech, unii-q7e9k52w7e, 2,6 dichlorophenol, ccris 2511, 2,6-dcp | |
OC1=C(Cl)C=CC=C1Cl | |
163.00 | |
CHEBI:28457 | |
99% |
65°C to 68°C | |
Phenol-like | |
MFCD00002176 | |
UN2020 | |
14,3073 | |
HOLHYSJJBXSLMV-UHFFFAOYSA-N | |
2,6-dichlorophenol | |
6899 | |
163 | |
2,6-Dichlorophenol |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROPHENOLS, SOLID
EINECSNumber : 201-761-3
RTECSNumber : SK8750000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only