Please login to your online account to display your discounted pricing

2,6-Dichlorophenethylamine, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 6-Dichlorophenethylamine
Name Note 99%
CAS 14573-23-0
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8 H9 Cl2 N
Formula Weight 190.07
MDL Number MFCD00060612
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC350790050

  SDS
thermo fisher scientific
350790050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 190.07
Quantity 5g
Color Clear colorless to slightly yellow
TSCA TSCA
Density 1.2800g/mL
CAS Max % 100.0
Infrared Spectrum Authentic
Refractive Index 1.5680 to 1.5700 (20°C, 589nm)
Chemical Name or Material 2, 6-Dichlorophenethylamine
Name Note 99%
Assay Percent Range 99%
Molecular Formula C8 H9 Cl2 N
MDL Number MFCD00060612
Packaging Glass bottle
CAS 14573-23-0
CAS Min % 98.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.