Please login to your online account to display your discounted pricing

2,6-Dichlorophenethyl alcohol, 96%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  35076-0050

 View more versions of this product

Catalog No. AC350760050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥95%
Physical Form Crystalline Powder, Crystals or Flakes
Formula Weight 191.06
Quantity 5g
Color White to slightly yellow
TSCA TSCA
Melting Point 74° to 79°C
CAS Min % 95.0
CAS Max % 100.0
Chemical Name or Material 2, 6-Dichlorophenethyl alcohol
Infrared Spectrum Authentic
Name Note 96%
Molecular Formula C8 H8 Cl2 O
MDL Number MFCD00800674
Packaging Glass bottle
CAS 30595-79-0
Assay Percent Range 96%

Specifications

Chemical Name or Material 2, 6-Dichlorophenethyl alcohol
Name Note 96%
CAS 30595-79-0
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C8 H8 Cl2 O
Formula Weight 191.06
MDL Number MFCD00800674