missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Dichlorophenethyl alcohol, 96%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 35076-0010

 View more versions of this product

Catalog No. AC350760010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2, 6-Dichlorophenethyl alcohol
74° to 79°C
96%
95.0
Authentic
≥95%
C8 H8 Cl2 O
MFCD00800674
White to slightly yellow
1g
30595-79-0
100.0
Crystalline Powder, Crystals or Flakes
96%
191.06
Glass bottle

Chemical Identifiers

30595-79-0
100.0
C8 H8 Cl2 O
MFCD00800674
≥95%
95.0
96%
191.06
2, 6-Dichlorophenethyl alcohol, 96%

Safety and Handling

SDS & Safety Documents

TSCA : TSCA