Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dichloronicotinic acid, 97%

$54.78 - $193.95

Chemical Identifiers

Chemical Name or Material 2,6-Dichloronicotinic acid
CAS 38496-18-3
Molecular Formula C6H3Cl2NO2
Formula Weight 192
MDL Number MFCD00075583
Beilstein 136114
Synonym 2,6-Dichloropyridine-3-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2000206

 
alfa aesar™
L2000206
5g Each for $193.95

AAL2000203

 
alfa aesar™
L2000203
1g Each for $54.78
Description & Specifications

Specifications

Formula Weight 192
Chemical Name or Material 2,6-Dichloronicotinic acid
MDL Number MFCD00075583
CAS 38496-18-3
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Molecular Formula C6H3Cl2NO2
Beilstein 136114
Melting Point 146° to 151°C
Synonym 2,6-Dichloropyridine-3-carboxylic acid
Health Hazard 1 Warning
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.