Learn More
2,6-Dichloroindophenol sodium salt hydrate, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1010722
Description
2,6-Dichloroindophenol sodium salt hydrate is suitable for vitamin C determination. It acts as a redox dye to investigate the efficiency of gold nanoparticles (Au-NP) for the enzymatic activity of glucose oxidase. It is also used to compose UVB specific dosimeter and to measure the rate of photosynthesis. It serves as a good reducing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,6-Dichloroindophenol sodium salt hydrate is suitable for vitamin C determination. It acts as a redox dye to investigate the efficiency of gold nanoparticles (Au-NP) for the enzymatic activity of glucose oxidase. It is also used to compose UVB specific dosimeter and to measure the rate of photosynthesis. It serves as a good reducing agent.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
2,6-Dichloroindophenol sodium salt hydrate | |
>300°C | |
MFCD00150014 | |
3641229 | |
14,3068 | |
Soluble in water. | |
[Na+].[O-]C1=C(Cl)C=C(C=C1Cl)N=C1C=CC(=O)C=C1 | |
290.07 | |
CHEBI:948 |
1266615-56-8 | |
C12H6Cl2NNaO2 | |
100 g | |
Light sensitive | |
2,6-dichloroindophenol sodium salt, 2,6-dichlorophenolindophenol sodium salt, tillman's reagent, tillman's reagenz, dichlorphenol-indophenolnatrium, 2,6-dichloroindophenol sodium, unii-kad7q8xo1y, sodium 2,6-dichloroindophenol, sodium 2,6-dichloroindophenolate, 2,6-dichloroindophenol, sodium salt | |
FHLDWQLHDYCXKI-UHFFFAOYSA-M | |
sodium;4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]phenolate | |
23697355 | |
290.08 (Anhydrous) |
Chemical Identifiers
1266615-56-8 | |
290.07 | |
FHLDWQLHDYCXKI-UHFFFAOYSA-M | |
23697355 | |
sodium;4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]phenolate |
C12H6Cl2NNaO2 | |
MFCD00150014 | |
2,6-dichloroindophenol sodium salt, 2,6-dichlorophenolindophenol sodium salt, tillman's reagent, tillman's reagenz, dichlorphenol-indophenolnatrium, 2,6-dichloroindophenol sodium, unii-kad7q8xo1y, sodium 2,6-dichloroindophenol, sodium 2,6-dichloroindophenolate, 2,6-dichloroindophenol, sodium salt | |
CHEBI:948 | |
[Na+].[O-]C1=C(Cl)C=C(C=C1Cl)N=C1C=CC(=O)C=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
H500
EINECSNumber : 210-640-4
RTECSNumber : GU5495000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only