missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dichloroindophenol sodium salt hydrate

Manufacturer:  Alfa Aesar™ A1010706

 View more versions of this product

Catalog No. AAA1010706



Description

Description

Applications
2,6-Dichloroindophenol sodium salt hydrate is suitable for vitamin C determination. It acts as a redox dye to investigate the efficiency of gold nanoparticles (Au-NP) for the enzymatic activity of glucose oxidase. It is also used to compose UVB specific dosimeter and to measure the rate of photosynthesis. It serves as a good reducing agent.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

2,6-Dichloroindophenol sodium salt hydrate
620-45-1
MFCD00150014
14,3068
2,6-Dichlorophenolindophenol sodium salt hydrate; Sodium 2,5-dichloroindophenoxide hydrate
C1=CC(=CC=C1N=C2C=C(C(=O)C(=C2)Cl)Cl)[O-].[Na+]
290.075
CHEBI:948
5g
C12H6Cl2NNaO2
3641229
>300°C
CVSUAFOWIXUYQA-UHFFFAOYSA-M
sodium;4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]phenolate
23697355
290.08 (Anhydrous)

Chemical Identifiers

620-45-1
290.075
CVSUAFOWIXUYQA-UHFFFAOYSA-M
23697355
sodium;4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]phenolate
C12H6Cl2NNaO2
MFCD00150014
2,6-Dichlorophenolindophenol sodium salt hydrate; Sodium 2,5-dichloroindophenoxide hydrate
CHEBI:948
C1=CC(=CC=C1N=C2C=C(C(=O)C(=C2)Cl)Cl)[O-].[Na+]
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : GU5495000

TSCA : TSCA

SDS Documents